Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD59 Mouse anti-Human, Clone: 2/24, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1759
Description
Detect CD59 using this Anti-CD59 Antibody, clone 2/24 validated for use in FC and immunofluoresence.
Specifications
CD59 | |
Monoclonal | |
Unconjugated | |
NM_000611.4; NM_203329.1; NM_203330.1; NM_203331.1 | |
Mouse | |
100 μg | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
2/24 | |
Purified from mouse ascitic fluid. The antibody is supplied in phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry. | |
CD59, MACIF, MIN2, MIN3, PROTECTIN, HRF-20, 16.3A5, MSK21, EJ16, HRF20, EL32, G344, MIC11, MIN1, Protectin, MAC-IP, MGC2354, EJ30, MIRL, p18-20 | |
Human erythrocytes | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction