Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD59 Mouse anti-Human, Unlabeled, Clone: P282 (H19), BD

Catalog No. BDB555761 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555761 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555761 Supplier BD Biosciences Supplier No. 555761
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The p282 (HI9) monoclonal antibody specifically binds to CD59, a 19 kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein, expressed on hematopoietic and non-hematopoietic cells. Because of its role in interaction with complement activated products, CD59 has been termed membrane-attack-complex-inhibitory factor (MACIF), homologus restriction factor (HRF20), membrane inhibitor of reactive lysis (MIRL) and Protectin. It inhibits the cytolytic activity of complement by binding to C8 and C9, thereby blocking the assembly of the membrane attack complex. CD59 also participates in spontaneous T-cell/erythrocyte adhesion, interacts with CD2 and plays a role in T-cell activation.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD59
Applications Flow Cytometry
Classification Monoclonal
Clone P282 (H19)
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Erythrocytes
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.