Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD59 Mouse anti-Human, FITC, Clone: 2/24, MilliporeSigma™

Catalog No. MAB1759FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1759FMI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1759FMI Supplier MilliporeSigma Supplier No. MAB1759F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD59 Antibody, clone 2/24, FITC conjugated detects level of CD59 and has been published and validated for use in FC and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD59
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 2/24
Conjugate FITC
Formulation Purified from mouse ascitic fluid. FITC Conjugated. The antibody is supplied in phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometr
Gene Accession No. NM_000611.4; NM_203329.1; NM_203330.1; NM_203331.1
Gene Symbols CD59, MACIF, MIN2, MIN3, PROTECTIN, HRF-20, 16.3A5, MSK21, EJ16, HRF20, EL32, G344, MIC11, MIN1, Protectin, MAC-IP, MGC2354, EJ30, MIRL, p18-20
Host Species Mouse
Immunogen Human erythrocytes
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.