Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD59 Mouse anti-Human, PE, Clone: P282 (H19), BD

Catalog No. BDB557141 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557141 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557141 Supplier BD Biosciences Supplier No. 557141
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone p282 reacts with the human form of the 19 kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein known as CD59, expressed on hematopoietic and non-hematopoietic cells. Clone p282 also cross-reacts with peripheral blood leukocytes of baboon and both rhesus and cynomolgus macaque monkeys. The distribution of leukocytes is similar to that observed with peripheral blood leukocytes from normal human donors, with all populations, lymphocytes, monocytes and granulocytes showing reactivity to p282.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD59
Applications Flow Cytometry
Classification Monoclonal
Clone P282 (H19)
Conjugate PE
Description HRF-20; MAC-inhibitory protein; MAC-IP; MACIF; MEM43; MIRL; Protectin; 1F5
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Erythrocytes
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Canine, Cynomolgus, Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.