Learn More
CD6 Mouse anti-Human, Alexa Fluor 647, Clone: M-T605, BD
Catalog No. BDB564260 Shop All BD Biosciences Products
Change view
Click to view available options
100 Tests; Alexa Fluor 647
Catalog No.
BDB564260 100 Tests; Alexa Fluor 647
1 options
This item is not returnable. View return policy
Catalog No. BDB564260 Supplier BD Biosciences Supplier No. 564260
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mouse Monoclonal Antibody
The M-T605 monoclonal antibody specifically binds to CD6. CD6 is a 100-130 kDa type I transmembrane glycoprotein present on more than 80% of mature T cells and a subset of B cells. CD6 is also weakly expressed on cortical thymocytes. CD6 binds to CD166 and reportedly plays roles in cellular adhesion, costimulation of T lymphocyte proliferation, and the regulation of lymphocyte apoptosis. The antibody is useful for investigation of CD6+ B-cell function.
TRUSTED_SUSTAINABILITY
WARNING: Reproductive Harm - www.P65Warnings.ca.gov