Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD6 Mouse anti-Human, Brilliant Violet 421, Clone: M-T605, BD

Catalog No. BDB564259 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564259 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564259 Supplier BD Biosciences Supplier No. 564259
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The M-T605 monoclonal antibody specifically binds to CD6. CD6 is a 100-130 kDa type I transmembrane glycoprotein present on more than 80% of mature T cells and a subset of B cells. CD6 is also weakly expressed on cortical thymocytes. CD6 binds to CD166 and reportedly plays roles in cellular adhesion, costimulation of T lymphocyte proliferation, and the regulation of lymphocyte apoptosis. The antibody is useful for investigation of CD6+ B-cell function.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD6
Applications Flow Cytometry
Classification Monoclonal
Clone M-T605
Conjugate Brilliant Violet 421
Description T12; TP120
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.