Learn More
CD6 Mouse anti-Rat, Unlabeled, Clone: OX-52, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550979
Description
The OX-52 antibody reacts with CD6, a 95-120–kDa antigen on T lymphocytes and thymocytes expressing αβ and γδ Tcell receptors. Rat NK cells express low levels of CD6 and are CD8a+, CD8b-, CD5-, and CD3-. In immunoprecipitation experiments, CD6 has been shown to co-precipitate with CD5 and to play a key role in CD5 tyrosine phosphorylation.

Specifications
CD6 | |
Monoclonal | |
0.5mg/mL | |
OX-52 Antigen, Pan-T Cells | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
OX-52 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
PVG-RT1c rat thoracic-duct leukocytes and splenocytes | |
0.1 mg | |
Immunology | |
Rat | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.