Learn More
CD6 Mouse anti-Human, FITC, Clone: M-T605, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555357
Description
The M-T605 monoclonal antibody specifically binds to CD6. CD6 is a 100-130 kDa type I transmembrane glycoprotein present on more than 80% of mature T cells and a subset of B cells. CD6 is also weakly expressed on cortical thymocytes. CD6 binds to CD166 and reportedly plays roles in cellular adhesion, costimulation of T lymphocyte proliferation, and the regulation of lymphocyte apoptosis. The antibody is useful for investigation of CD6+ B-cell function.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
CD6 | |
Monoclonal | |
FITC | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
M-T605 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.