Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD6 Mouse anti-Rat, FITC, Clone: OX-52, BD

Catalog No. BDB554904 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554904 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554904 Supplier BD Biosciences Supplier No. 554904
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The OX-52 antibody reacts with CD6, a 95-120–kDa antigen on T lymphocytes and thymocytes expressing αβ and γδ Tcell receptors. Rat NK cells express low levels of CD6 and are CD8a+, CD8b-, CD5-, and CD3-. In immunoprecipitation experiments, CD6 has been shown to co-precipitate with CD5 and to play a key role in CD5 tyrosine phosphorylation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD6
Applications Flow Cytometry
Classification Monoclonal
Clone OX-52
Concentration 0.5mg/mL
Conjugate FITC
Description OX-52 Antigen, Pan-T Cells
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen PVG-RT1c rat thoracic-duct leukocytes and splenocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.