Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD61 Mouse anti-Human, Brilliant Violet 510, Clone: VI-PL2, BD

Catalog No. BDB563303 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563303 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563303 Supplier BD Biosciences Supplier No. 563303
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The VI-PL2 monoclonal antibody specifically binds to CD61. CD61 is a 105 kDa transmembrane glycoprotein that is also known as integrin β3 and platelet glycoprotein IIIa (GPIIIa or GP3A). It is expressed on platelets, megakaryocytes, osteoclasts and endothelia. Integrin β3 associates with gpIIa (CD41) to form the CD41/CD61 complex which mediates platelet adhesion and aggregation. CD61 also associates with CD51 to form the CD51/CD61 complex (vitronectin receptor). CD61 appears to bind to fibrinogen, fibronectin, vWF, vitronectin, and thrombospondin to mediate cell adhesion.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD61
Applications Flow Cytometry
Classification Monoclonal
Clone VI-PL2
Conjugate Brilliant Violet 510
Description Integrin β3; Integrin beta-3; GP3A; GPIIIa; ITGB3; ITB3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.