Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD61 Hamster anti-Mouse, Unlabeled, Clone: 2C9.G2, BD

Catalog No. BDB550541 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550541 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550541 Supplier BD Biosciences Supplier No. 550541
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The 2C9.G2 antibody reacts with the integrin β;3 chain (CD61), which associates with the integrin α;v chain (CD51) to form the vitronectin receptor and with the α;IIb chain (CD41) to form the gpIIb/IIIa complex. Both receptors mediate adhesion to fibronectin, fibrinogen, vitronectin, thrombospondin, and von Willebrand factor. Leukocyte-endothelial adhesion is also mediated by the binding of α;vβ;3 integrin or vitronectin receptor to CD31 (PECAM-1). In addition, interaction of the α;vβ;3 integrin with its ligands regulates the L-type Ca2+ channel in vascular smooth muscle cells, possibly mediating vasodilatory responses to injury. Soluble and insoluble 2C9.G2 mAb mimics the effect of the natural ligands in smooth muscle cells from rat cremaster arterioles. Furthermore, osteopontin, also named Eta-1, is a cytokine that binds to α;vβ;3. CD61 is expressed on platelets, activated T lymphocytes, polymorphonuclear granulocytes, and blastocysts. Cross-reactivity of mAb 2C9.G2 to rat mast cells and platelets has been observed by flow cytometric analysis. mAb 2C9.G2 has been demonstrated to block binding of rat and mouse cells to fibronectin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD61
Applications Flow Cytometry
Classification Monoclonal
Clone 2C9.G2
Concentration 250μg/mL
Conjugate Unconjugated
Description Integrin β3 chain
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse T-cell Hybridoma 2B4 Vitronectin Receptor
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.