Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD61 Mouse anti-Rat, Unlabeled, Clone: F11, BD

Catalog No. BDB554951 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554951 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554951 Supplier BD Biosciences Supplier No. 554951
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The F11 antibody reacts with the integrin β3 chain (CD61), which associates with the integrin αv chain (CD51), to form the vitronectin receptor found on endothelial cells, myeloid cells, and osteoclasts, and with the αIIb chain (CD41) on platelets and megakaryocytes. Both receptors mediate adhesion to fibronectin, fibrinogen, vitronectin, thrombospondin, and von Willebrand factor. F11 mAb has been reported to block the in vitro attachment of osteoclasts to several ligands. Weak reactivity with human osteoclasts, megakaryocytes, and platelets has also been observed. Other reported applications include immunoprecipitation, in vitro blocking, and immunohistochemical staining (IHC) of acetone-fixed frozen and zinc-fixed paraffin-embedded sections. IHC of formalin-fixed paraffin-embedded sections is not recommended.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD61
Applications Flow Cytometry
Classification Monoclonal
Clone F11
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Integrin β3 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat bone cell suspensions
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.