Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD61 (Integrin β3 chain) Mouse anti-Human, PerCP, Clone: RUU-PL7F12, BD

Catalog No. BDB340506 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340506 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340506 Supplier BD Biosciences Supplier No. 340506
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD61, clone RUU-PL 7F12, is derived from hybridization of mouse P3X63.Ag8.653 myeloma cells with spleen cells from BALB/c mice immunized with purified platelet membrane glycoproteins.

CD61 recognizes an Mr 110-kdalton (kDa) protein, also known as gpIIIa, the common β-subunit (integrin β3-chain) of the gpIIb/IIIa complex and the vitronectin receptor (VNR).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD61 (Integrin β3 chain)
Applications Flow Cytometry
Classification Monoclonal
Clone RUU-PL7F12
Conjugate PerCP
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Purified platelet membrane blycoproteins
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Form Purified
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.