Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62E Rat anti-Mouse, Biotin, Clone: 10E9.6, BD

Catalog No. BDB553750 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553750 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553750 Supplier BD Biosciences Supplier No. 553750
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 10E9.6 antibody reacts with the 97-110 kDa cell surface glycoprotein E-selectin (CD62E), also known as endothelial-leukocyte adhesion molecule-1 (ELAM-1), which is expressed on endotoxin- or cytokine-stimulated mouse endothelial cells. A suspension of TNFα stimulated mouse brain capillary endothelioma cells, from the cell line bEnd.3, was used as the immunogen. The epitope recognized by mAb 10E9.6 has been mapped to the first and/or second complement regulatory protein repeat domains of E-selectin. The 10E9.6 antibody has been reported to block binding of a monocyte cell line to E-selectin in vitro and to block neutrophil migration in BALB/c, but not C57BL/6 mice. It has no effect on leukocyte rolling in TNFα-treated mouse venules or on in vitro adhesion of myeloid cells to E-selectin. Studies have demonstrated that Cutaneous Lymphocyte Antigen (CLA), recognized by mAb HECA-452 (Cat. no. 555946), may be a ligand for CD62E.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62E
Applications Flow Cytometry
Classification Monoclonal
Clone 10E9.6
Concentration 0.5mg/mL
Conjugate Biotin
Description E-selectin, ELAM-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse brain capillary endothelioma bEnd.3 (TNFα-stimulated) Cell Line
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.