Learn More
CD62E Mouse anti-Human, PE, Clone: 68-5H11, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 551145
Description
The 68-5H11 monoclonal antibody specifically binds to CD62E. This adhesion molecule is a 97-115 kDa type I transmembrane glycoprotein that is encoded by the SELE gene. CD62E is also known as E-selectin, Endothelial-leukocyte adhesion molecule-1 (ELAM-1), and Leukocyte endothelial cell adhesion molecule 2 (LECAM-2). CD62E is minimally expressed by unstimulated endothelium. Activated endothelial cells upregulate surface CD62E expression in response to various activators including inflammatory cytokines such as Interleukin-1 and Tumor Necrosis Factor. CD62E plays a role in leucocyte extravasation by promoting leucocyte rolling on activated endothelial cells during inflammation. CD62E may also play a role in the metastasis of certain tumor cells. The adhesion between endothelial CD62E molecules and a carbohydrate ligand on neutrophils is inhibited by the mAb 68-5H11.

Specifications
CD62E | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
68-5H11 | |
SELE; E-selectin; Selectin E; ELAM; ELAM1; ESEL; LECAM2 | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.