Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62L Mouse anti-Human, BB515, Clone: SK11, BD

Catalog No. BDB565037 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565037 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565037 Supplier BD Biosciences Supplier No. 565037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SK11 monoclonal antibody specifically binds to CD62L which is also known as, L-selectin, Leukocyte-endothelial cell adhesion molecule 1 (LECAM1/LECAM-1), Leukocyte adhesion molecule 1 (LAM1/LAM-1), Lymph node homing receptor (LNHR), Leu-8, or TQ1. CD62L is an ∽80 kDa type I transmembrane glycoprotein that belongs to the Selectin/LECAM family. CD62L is differentially expressed on T cells, B cells, monocytes, granulocytes, and a subset of NK cells. The CD62L molecule is the human homolog of the mouse lymph node homing receptor, MEL-14. CD62L plays a role in leucocyte binding to inflamed endothelium and extravasation, as well as mediating lymphocyte homing into peripheral lymphoid tissues through high endothelial postcapillary venules. Soluble CD62L can result from the proteolytic cleavage of cell surface CD62L during cellular activation or inflammation.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62L
Applications Flow Cytometry
Classification Monoclonal
Clone SK11
Conjugate Brilliant Blue 515 (BB515)
Description L-selectin; LECAM-1; SELL; LNHR; LSEL; LAM-1; LEU8; PLNHR; TQ-1; MEL-14
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Human Peripheral Blood T Lymphocytes
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.