Learn More
Description
The SK11 monoclonal antibody specifically binds to CD62L which is also known as, L-selectin, Leukocyte-endothelial cell adhesion molecule 1 (LECAM1/LECAM-1), Leukocyte adhesion molecule 1 (LAM1/LAM-1), Lymph node homing receptor (LNHR), Leu-8, or TQ1. CD62L is an ∽80 kDa type I transmembrane glycoprotein that belongs to the Selectin/LECAM family. CD62L is differentially expressed on T cells, B cells, monocytes, granulocytes, and a subset of NK cells. The CD62L molecule is the human homolog of the mouse lymph node homing receptor, MEL-14. CD62L plays a role in leucocyte binding to inflamed endothelium and extravasation, as well as mediating lymphocyte homing into peripheral lymphoid tissues through high endothelial postcapillary venules. Soluble CD62L can result from the proteolytic cleavage of cell surface CD62L during cellular activation or inflammation.
The antibody was conjugated to BD Horizon BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
Specifications
| Antigen | CD62L |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SK11 |
| Conjugate | Brilliant Violet 711 |
| Description | L-selectin; LECAM-1; SELL; LNHR; LSEL; LAM-1; LEU8; PLNHR; TQ-1; MEL-14 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human Peripheral Blood T Lymphocytes |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.