Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62P Mouse anti-Human, PE, Clone: AC1.2, BD

Catalog No. BDB550561 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550561 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550561 Supplier BD Biosciences Supplier No. 550561
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the human form of the 140kDa membrane glycoprotein, P-selectin, formerly known as platelet activation-dependent granule external membrane protein (PADGEM), or granule membrane protein (GMP-140). P-selectin is stored in the α-granules of platelets and the Weibel-Palade bodies of endothelial cells, and is rapidly transported to the plasma membrane upon activation. P-selectin is thought to mediate the initial adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses, and of activated platelets to neutrophils and monocytes in hemostasis. This antibody is a non-blocking monoclonal antibody and is suitable for immunohistochemical staining of formalin-fixed, paraffin embedded, tissue sections. Clone AC1.2 also cross reacts with activated platelets of baboon, and both rhesus and cynomolgus macaque monkeys. The reactivity on resting and activated platelets is similar to that observed on resting and activated platelets of human donors.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62P
Applications Flow Cytometry
Classification Monoclonal
Clone AC1.2
Conjugate PE
Description P-Selectin
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.