Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62P (P-selectin) Mouse anti-Human, PE, Clone: AC1.2, BD

Catalog No. BDB348107 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB348107 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB348107 Supplier BD Biosciences Supplier No. 348107
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD62P, clone AC1.2, is derived from hybridization of mouse NS-1 cells with spleen cells of BALB/c mice immunized with activated platelets.

The CD62P antigen, also known as platelet activation-dependent granule-external membrane (PADGEM) protein or granule membrane protein (GMP-140), is a 140-kilodalton (kd) single chain polypeptide. The CD62P antigen is a member of the selectin family of adhesion molecules and mediates the adhesion of activated platelets to neutrophils and monocytes in hemostasis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62P (P-selectin)
Applications Flow Cytometry
Classification Monoclonal
Clone AC1.2
Concentration 1.5μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.