Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63 Mouse anti-Rat, Unlabeled, Clone: AD1, BD

Catalog No. BDB551458 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551458 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551458 Supplier BD Biosciences Supplier No. 551458
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AD1 antibody reacts with a 50-60-kDa protein that is a homolog (87%) ofhuman CD63 (melanomaassociated antigen ME491). CD63, a highly glycosylated type-III lysosomal glycoprotein, is a member of the tetraspan transmembrane 4 superfamily (TM4SF) that also includes CD9, CD37, and CD53. In the rat, CD63 is expressed on mast cells and platelets. CD63 is also found on rat embryonic fibroblasts and hepatic cell, along with cultured cell lines of diverse origin (eg, liver, lung, and heart). Expression is upregulated with culture. CD63 is expressed in close proximity to the FcεRI on rat mast cells and has been shown to partially inhibit IgE-mediated histamine release. The function of CD63 has not been fully characterized.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD63
Applications Flow Cytometry
Classification Monoclonal
Clone AD1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description ME491
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat basophilic leukemia cell line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.