Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD63, ; Clone: NVG-2; BD

Catalog No. BDB564221 Shop All BD Biosciences Products
Change view
Click to view available options
50μg;
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BDB564221 50μg;
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564221 Supplier BD Biosciences Supplier No. 564221
Add to Cart
Add to Cart

Applications:

Flow cytometry, Immunofluorescence, Bioimaging, Intracellular staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD63
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone NVG-2
Concentration 0.5mg/mL
Host Species Rat
Immunogen Mouse Eosinophils
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4deg.C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.