Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63 Mouse anti-Human, Clone: RFAC4, MilliporeSigma™

Catalog No. CBL553MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL553MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL553MI Supplier MilliporeSigma Supplier No. CBL553
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD63 Antibody, clone RFAC4 is an antibody against CD63 for use in flow cytometry, immunoprecipitation, western blot, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD63
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RFAC4
Conjugate Unconjugated
Formulation The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular appli
Gene Accession No. NM_001040034.1; NM_001780.4
Gene Symbols CD63, TSPAN30, ME491, Tetraspanin-30, granulophysin, LAMP-3, tetraspanin-30, Granulophysin, MLA1, OMA81H, Lamp-3
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.