Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD64 a and b Alloantigens Mouse anti-Mouse, Alexa Fluor 647, Clone: X54-5/7.1, BD

Catalog No. BDB558539 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558539 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558539 Supplier BD Biosciences Supplier No. 558539
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal allo antibody X54-5/7.1 reacts with FcγRI (CD64) encoded by the more common Fcgr1a and Fcgr1b alleles. The alloantigens generated by the Fcgr1a and Fcgr1b alleles, have been confirmed positive in mouse strains BALB/c and C57BL/6 and reported positive in strains 129, A, AKR, ALR, BUB, C3H, C57BL/10, C57BLKS, C57BR, C58, CBA, CE, DBA/2, HRS, MRL, NON, NZB, NZO, NZW, PL, SJL, ST, SWR. The a and b alloantigens have been reported negative in mouse strains ABH, NOD. CD64, a key receptor in the development of immune responses, has a dual role as a low affinity receptor for IgG3 and high affinity receptor for IgG2a linking innate and adaptive immunities. CD64 mediates endocytosis, phagocytosis, antibody-dependent cellular toxicity, cytokine release and superoxide generation. CD64 is expressed largely on macrophages and dendritic cells. For more information regarding clone X54-5/7.1 and the alloantigens it recognizes, please refer to the reference by Tan et al listed below.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD64 a and b Alloantigens
Applications Flow Cytometry
Classification Monoclonal
Clone X54-5/7.1
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description FcgRI
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse CD64 a Alloantigen
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.