Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD64 Mouse anti-Human, PE, Clone: 10.1, BD

Catalog No. BDB644385 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB644385 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB644385 Supplier BD Biosciences Supplier No. 644385
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD64, clone 10.1, is derived from a fusion of Sp2/0-Ag14 cells with spleen cells from a BALB/c mouse that was immunized first with 2 x 107 rheumatoid synovial fluid cells and on subsequent occasions with 1.5 x 107 fibronectin-purified human monocytes obtained from pools of blood group–matched donors.

CD64 recognizes the 72-kilodalton (kDa) human FcγRI that can bind monomeric IgG.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD64
Applications Flow Cytometry
Classification Monoclonal
Clone 10.1
Concentration 25μg/mL
Conjugate PE
Formulation Phosphate buffered saline with BSA and 0.1% sodium azide.
Host Species Mouse
Immunogen Human Rheumatoid synovial fluid cells and fibronectin-purified monocytes
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.