Learn More
CD66 Mouse anti-Human, Unlabeled, Clone: B1.1/CD66, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 551354
Description
The B1.1 monoclonal antibody binds to several glycosylphosphatidylinositol-anchored glycoproteins present on granulocytes and epithelial cells. As reported in the Sixth International Workshop and Conference on Human Leucocyte Differentiation Antigens, the B1.1 antibody recognized CD66a (CEACAM1), CD66c (CEACAM6), CD66d (CEACAM3) and CD66e (. These molecules exhibit both homophilic and heterophilic adhesion. CEA family members may be involved in transmembrane signaling and activation of neutrophils.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
CD66 | |
Monoclonal | |
0.5mg/mL | |
CEA, carcinoembryonic antigen, CD66a, CD66c, CD66d, CD66e | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
B1.1/CD66 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Breast Carcinoma Cells | |
0.1 mg | |
Immunology | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.