Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD66abce Mouse anti-Human, FITC, Clone: Kat4c, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma FCMAB204F
Description
Detect CD66abce using this Milli-Mark™ Anti-CD66abce-FITC Antibody, clone Kat4c validated for use in flow cytometry.
Specifications
CD66abce | |
Monoclonal | |
FITC | |
NP_001020083.1 | |
Protein A purfied | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. Protect from light. | |
IgG1 κ |
Flow Cytometry | |
Kat4c | |
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2. | |
Mouse | |
100 Tests | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction