Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD66b Mouse anti-Human, Alexa Fluor 647, Clone: G10F5, BD

Catalog No. BDB561645 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561645 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561645 Supplier BD Biosciences Supplier No. 561645
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G10F5 monoclonal antibody specifically binds to CD66b, also known as Carcinoembryonic antigen-related cell adhesion molecule 8 (CEACAM8). CD66b is a glycosylphosphatidylinositol (GPI) linked protein with amolecular weight of 100 kDa expressed on granulocytes. This molecule was previously clustered as CD67 in the Fourth Human Leucocyte DifferentiationAntigen (HLDA) Workshop and renamed CD66b in the Fifth HLDA Workshop. CD66b is a member of the carcinoembryonic antigen (CEA)-like glycoprotein family present on granulocytes and referred to as non-specific crossreacting antigens (NCA). Granulocyte activation induced with soluble stimulators (calcium ionophore, phorbol myristate acetate, N-formylmethionyl- leucyl-phenylalanine) results in release and increased expression of NCA. Findings suggest that these molecules may play a role in phagocytosis, chemotaxis and adherence.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD66b
Applications Flow Cytometry
Classification Monoclonal
Clone G10F5
Conjugate Alexa Fluor 647
Description CEACAM8; CGM6; NCA-95
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Granulocytes
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.