Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD66b Mouse anti-Human, BB515, Clone: G10F5, BD

Catalog No. BDB564679 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564679 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564679 Supplier BD Biosciences Supplier No. 564679
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G10F5 monoclonal antibody specifically binds to CD66b, also known as Carcinoembryonic antigen-related cell adhesion molecule 8 (CEACAM8). CD66b is a glycosylphosphatidylinositol (GPI) linked protein with a molecular weight of 100 kDa expressed on granulocytes. This molecule was previously clustered as CD67 in the Fourth Human Leucocyte Differentiation Antigen (HLDA) Workshop and renamed CD66b in the Fifth HLDA Workshop. CD66b is a member of the carcinoembryonic antigen (CEA)-like glycoprotein family present on granulocytes and referred to as non-specific crossreacting antigens (NCA). Granulocyte activation induced with soluble stimulators (calcium ionophore, phorbol myristate acetate, N-formylmethionyl- leucyl-phenylalanine) results in release and increased expression of NCA. Findings suggest that these molecules may play a role in phagocytosis, chemotaxis and adherence.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (eg, 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD66b
Applications Flow Cytometry
Classification Monoclonal
Clone G10F5
Conjugate Brilliant Blue 515 (BB515)
Description CEACAM8; CGM6; NCA-95
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Human Granulocytes
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.