Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD66c Mouse anti-Human, PE, Clone: B6.2/CD66C, BD

Catalog No. BDB551478 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551478 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551478 Supplier BD Biosciences Supplier No. 551478
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with two glycosylphosphatidylinositol-anchored glycoprotein present on granulocytes. Antibody B6.2 was studied as recognizing CD66c in the VI Human Leukocyte Differentiation Antigen workshop. CD66 antigens also known as the carcinoembryonic antigen (CEA) family of molecules, are closely related to the immunoglobulin super family of glycoproteins. Studies on CD66 molecules suggest a potential adhesion function in vivo. These molecules exhibit both homophilic and heterophilic adhesion. CEA family members may be involved intransmembrane signalling and activation of neutrophils. This clone has been found to be N-terminal domain reactive, reacted preferentially with the native protein and were conformationally dependent.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD66c
Applications Flow Cytometry
Classification Monoclonal
Clone B6.2/CD66C
Conjugate PE
Description CEA, carcinoembryonic antigen
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.