Learn More
CD66f Mouse anti-Human, Unlabeled, Clone: IID10, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559587
Description
Reacts with placenta-specific glycoprotein, SP-1, a 54-72 kDa molecule also known as pregnancy-specific glycoprotein (PSG). CD66f belongs to the carcinoembryonic family of proteins. It is produced in placental syncytiotrophoblasts, fetal liver, and myeloid cell lines. Its role in immune regulation and protection of fetus from maternal immune system has not been fully elucidated. This reagent is suitable for immunohistochemical staining of formalin-fixed, paraffin-embedded tissue sections.

Specifications
CD66f | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Immunohistochemistry (Frozen) | |
IID10 | |
Unconjugated | |
Mouse | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.