Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD68 Mouse anti-Human, Alexa Fluor 647, Clone: Y1/82A, BD

Catalog No. BDB562111 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562111 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562111 Supplier BD Biosciences Supplier No. 562111
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Y1/82A monoclonal antibody specifically binds to CD68 which is also known as, Scavenger receptor class D, member 1 (SCARD1), Macrosialin, or GP110. CD68 is a cell surface 110 kDa-type I-transmembrane glycoprotein that is primarily expressed in cytoplasmic granules of monocytes, macrophages, dendritic cells, granulocytes, myeloid progenitor cells and, reportedly, a subset of CD34-positive hemopoietic bone marrow progenitor cells. CD68 belongs to the sialomucin family and serves as a scavenger receptor that can bind and internalize oxidized low density lipoproteins (LDL). This antibody is useful in studies of myeloid cell development and function.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD68
Applications Flow Cytometry
Classification Monoclonal
Clone Y1/82A
Conjugate Alexa Fluor 647
Description GP110; Macrosialin; SCARD1; Scavenger receptor class D, member 1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen PHA-stimulated Human PBMC
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.