Learn More
CD69 Mouse anti-Human, APC, Clone: FN50, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555533
Description
Reacts with a 28/34kDa dimeric glycoprotein expressed early during activation of lymphocytes and monocytes. FN50 monoclonal antibody labels NK cells and most lymphocytes of the follicular mantle and perifollicular/interfollicular zone as well as intragerminal center T cells of lymph nodes and tonsils.

Specifications
CD69 | |
Monoclonal | |
APC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
FN50 | |
AIM; CLEC2C; EA1; GP32/28; Leu23; MLR-3; VEA; BL-AC/P26 | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.