Learn More
Description
Host Species: Hamster
Clone H1.2F3
Isotype IgG1, λ3
Species Reactivity Ms
Immunogen Mouse Dendritic Epidermal T Cell Line Y245
Flow Cytometry
Specifications
Specifications
| Antigen | CD69 |
| Classification | Monoclonal |
| Clone | H1.2F3 |
| Concentration | 0.2mg/mL |
| Host Species | Hamster |
| Immunogen | Mouse Dendritic Epidermal T Cell Line Y245 |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.