Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69 Mouse anti-Human, PE, Clone: L78, BD

Catalog No. BDB341652 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB341652 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB341652 Supplier BD Biosciences Supplier No. 341652
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD69, clone L78, is derived from hybridization of mouse Sp2/0-Ag14 myeloma cells with lymph node cells from BALB/c mice immunized with a CD8+ alloantigen–directed cytotoxic T-lymphocyte (CTL) cell line.

CD69 recognizes a very early human activation antigen that is a disulfide-bonded homodimer consisting of Mr 60-kilodalton (kd) polypeptides with one or two N-linked oligosaccharides.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD69
Applications Flow Cytometry
Classification Monoclonal
Clone L78
Concentration 3.0μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen CD8
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.