Learn More
CD7 Mouse anti-Human, APC-H7, Clone: M-T701, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564020
Description
The M-T701 monoclonal antibody specifically binds to CD7. CD7 is a 40 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin superfamily. The CD7 antigen is also known as Leu-9, TP41, Tp40, GP40, and T-cell leukemia antigen. It is expressed on thymocytes, T cells, pre-B cells, and NK cells. CD7 is present in reduced density on monocytic cells and cell lines. Functional studies demonstrate that crosslinking of the CD7 can induce transmembrane calcium flux in T cells.

Specifications
CD7 | |
Monoclonal | |
APC-H7 | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
P-CLL and Jurkat Cells | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
M-T701 | |
GP40; LEU-9; T-cell leukemia antigen; Tp40; TP41 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.