Learn More
CD7 Mouse anti-Human, FITC, Clone: M-T701, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 340737
Description
The CD7 antibody, clone M-T701, is derived from the hybridization of mouse P3-X63-Ag8.653 cells with spleen cells from BALB/c mice immunized with P-CLL and Jurkat cells.
The CD7 antibody recognizes a 40-kilodalton (kDa) type I transmembrane glycoprotein that is a member of the immunoglobulin superfamily (IgSF). The CD7 antigen is also known as Leu-9, TP41, Tp40, GP40, and T-cell leukemia antigen.

Specifications
CD7 | |
Monoclonal | |
12.5μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
P-CLL and Jurkat Cells | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
M-T701 | |
FITC | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.