Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD7 Mouse anti-Human, PerCP-Cy5.5, Clone: M-T701, BD

Catalog No. BDB561602 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561602 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561602 Supplier BD Biosciences Supplier No. 561602
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The M-T701 monoclonal antibody specifically binds to CD7. CD7 is a 40 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin superfamily. The CD7 antigen is also known as Leu-9, TP41, Tp40, GP40, and T-cell leukemia antigen. It is expressed on thymocytes, T cells, pre-B cells, and NK cells. CD7 is present in reduced density on monocytic cells and cell lines. Functional studies demonstrate that crosslinking of the CD7 can induce transmembrane calcium flux in T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD7
Applications Flow Cytometry
Classification Monoclonal
Clone M-T701
Conjugate PerCP-Cyanine5.5
Description GP40; LEU-9; T-cell leukemia antigen; Tp40; TP41
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen P-CLL and Jurkat Cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.