Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD71 Rat anti-Mouse, Brilliant Violet 605, Clone: C2, BD

Catalog No. BDB563013 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563013 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563013 Supplier BD Biosciences Supplier No. 563013
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The C2 monoclonal antibody specifically binds to CD71, the transferrin receptor. CD71 is a disulfide-linked homodimer of 95-kDa subunits. CD71 mediates one of the cellular mechanisms for iron uptake, and its expression is regulated according to the cell’s iron requirements. It is expressed at high levels on developing erythroid cells, and it is upregulated after mitogenic activation of B or T lymphocytes. The C2 monoclonal antibody selectivity inhibits some types of T- and B-cell activation by down-regulation of transferrin receptor expression, but it does not block binding of transferrin.

This antibody is conjugated to BD Horizon BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD71
Applications Flow Cytometry
Classification Monoclonal
Clone C2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Description Transferrin Receptor; TR; TfR; TfR1; Tfrc; Trfr; Mtvr-1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse cell line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.