Learn More
CD71 Mouse anti-Rat, PE, Clone: OX-26, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 554891
Description
The OX-26 antibody reacts with transferrin receptor found on proliferating cell and brain endothelium. It does not block transferrin binding.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
CD71 | |
Monoclonal | |
0.2mg/mL | |
Transferrin Receptor | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
OX-26 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
PHA-activated PVG rat lymph node cells | |
0.2 mg | |
Immunology | |
Rat | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.