Learn More
CD71 (Transferrin Receptor) Mouse anti-Human, Unlabeled, Clone: L01.1, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 347510
Description
The CD71 antibody, clone L01.1, is derived from the hybridization of P3-X63- Ag8.653 mouse myeloma cells with spleen cells from BALB/c mice immunized with blast cells from pokeweed mitogen–stimulated peripheral blood cells.
The CD71 antibody is specific for the human transferrin receptor, with a molecular weight of 190 kilodaltons (kDa), 90-kDa reduced.

Specifications
CD71 (Transferrin Receptor) | |
Monoclonal | |
100μg/mL | |
Transferrin Receptor | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
L01.1 | |
Unconjugated | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Blast cells from pokeweed mitogen-stimulated peripheral blood cells | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.