Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD72 a, b, d Alloantigens Mouse anti-Mouse, FITC, Clone: K10.6, BD

Catalog No. BDB550966 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550966 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550966 Supplier BD Biosciences Supplier No. 550966
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The K10.6 monoclonal antibody specifically recognizes Lyb-2.1, Lyb-2.2, and Lyb-2.4 (CD72 a, b, and d alloantigens, respectively). CD72 is a 45-kDa type-II membrane protein, containing a C-type lectin-like domain and ITIM and ITIM-like sequences in the cytoplasmic tail. CD72 is expressed at all stages of B-lymphocyte development except plasma cells, and it has been shown to negatively regulate B-cell receptor signaling. Analysis of CD72-deficient mice supports these results and shows that CD72 is involved in B-cell development. CD72-stimulated B cells show a transient association of CD19 with CD72, as well as an increase in Tyr-phosphorylation of CD19. CD72 is reported to be a ligand for CD5, although this is controversial. It is also reported to be a ligand for CD100 (Sema4D). The CD72 alloantigens Lyb-2.1 (originally identified as Ly-m19.2), Lyb-2.2 (originally identified as Ly-32.2), Lyb-2.3, and Lyb-2.4 are encoded by the Cd72[a], Cd72[b], Cd72[c], and Cd72[d] alleles, respectively. Lyb-2.1 is expressed on B lymphocytes of CBA/J, C3H/Bi, C57BR, C57L, C58, DBA/1, DBA/2, and SWR strains. Lyb-2.2 is expressed on B lymphocytes, a subset of peripheral T cells, and activated T lymphocytes in A, BALB/c, CBA/H, C3H/He, C57BL, PL, and 129 strains. Lyb-2.4 is expressed on a subset of splenocytes of the STS/A strain. K10.6 antibody does not react with Lyb-2.3 (AKR and SJL strains) nor with non-lymphoid tissues. Five serological specificities of CD72 alloantigens have been described and the nomenclature CD72.1, CD72.2, CD72.3, CD72.4, and CD72.5 proposed, which does not correspond to the names of the alleles. Some authors have referred to the specificity of mAb K10.6 as CD72.4.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD72 a, b, d Alloantigens
Applications Flow Cytometry
Classification Monoclonal
Clone K10.6
Concentration 0.5mg/mL
Conjugate FITC
Description Lyb-2.1, Lyb-2.2, and Lyb-2.4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen (C57BL/6, DBA/2)F1 hybrid mouse pre-B-cell leukemia line 70Z/3
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.