Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD73 Rat anti-Mouse, PE, Clone: TY/23, BD

Catalog No. BDB550741 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550741 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550741 Supplier BD Biosciences Supplier No. 550741
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The TY/23 antibody reacts with CD73 or Ecto-5'-nucleotidase (5'-NT), a 69 kDa GPI-anchored cell-surface protein with enzymatic and signal transduction activities. 5'-NT catalyzes the dephosphorylation of extracellular nucleoside 5' monophosphates into a form which can enter cells to meet their metabolic needs. It also regulates the concentration of extracellular adenosine, which initiates a variety of physiological responses through the adenosine receptors in many tissues. CD73 expression appears to be developmentally regulated on leukocytes. In the bone marrow, it is found on most CD11b+ myeloid cells and very few CD19+ cells of the B-lymphocyte lineage. It is not found on CD11b+ cells in the periphery nor on marrow-derived GMCSF-stimulated dendritic cells. Some peripheral B lymphocytes express CD73, with higher levels on isotype-switched B cells. The few thymocytes which have detectable surface CD73 are of the CD4-CD8- (double negative) and the single-positive populations. In the peripheral lymphoid organs, significant subpopulations of the CD4+ and CD8+ T lymphocytes express CD73, with variation in the percentages of CD73-bearing T cells observed among inbred mouse strains. In the thymus and peripheral lymphoid organs, CD73 is found on endothelia and stromal cells. CD73 has also been detected on bone marrow and thymic epithelial cell lines, kidney glomeruli and proximal-tubule epithelial cells, and liver endothelial cells and hepatocytes. The TY/23 mAb inhibits the enzymatic activity of CD73. Although soluble TY/23 mAb by itself does not affect T lymphocyte proliferation, it is an effective co-stimulator with PMA, but not with Concanavalin-A or plate-bound anti-CD3ε mAb 145-2C11.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD73
Applications Flow Cytometry
Classification Monoclonal
Clone TY/23
Concentration 0.2mg/mL
Conjugate PE
Description Ecto-5'-nucleotidase
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen BALB/c mouse splenocytes and CHO cells transfected with the Mouse CD73
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.