Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD74 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7478
This item is not returnable.
View return policy
Description
CD74 Monoclonal specifically detects CD74 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| CD74 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P04441 | |
| CD74 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD74 Gln56-Cys215 Accession # P04441 | |
| 100 μg | |
| Primary | |
| Detects mouse CD74 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CD74 is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 829706 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD74, CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, CLIP, DHLAG, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, INVG34, MHC HLA-DR gamma chain, p33 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 972 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction