Learn More
CD74 Mouse anti-Human, Unlabeled, Clone: M-B741, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555538
Description
Reacts with cell -surface MHC class II-associated invariant chain molecule (41, 35, 33 kD). CD74 is moderately expressed on B cells, weakly on monocytes and macrophages. Other reactive cells include Langerhans cells, dendritic cells, a subset of activated T cells and thymic epithelium. CD74 is postulated to play a role in antigen processing and presentation through intracellular transport and expression of class II molecules on the cell surface.

Specifications
CD74 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
M-B741 | |
Unconjugated | |
Mouse | |
0.1 mg | |
Immunology | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.