Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD75 Mouse anti-Human, FITC, Clone: LN1, BD

Catalog No. BDB555654 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555654 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555654 Supplier BD Biosciences Supplier No. 555654
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with CD75, a carbohydrate determinant generated by the action of intracellular α2,6 sialyltransferase, expressed on mature B cells. In the lymph node, strong reactivity has been detected on germinal center B cells and weaker expression observed in mantle zone B cells. Evidence suggests that CD75 plays a role in intercellular adhesion and is a ligand for CD22.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD75
Applications Flow Cytometry
Classification Monoclonal
Clone LN1
Conjugate FITC
Description CDw75
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.