Learn More
CD77 Mouse anti-Human, Brilliant Violet 510, Clone: 5B5, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 563630
Description
The 5B5 monoclonal antibody specifically binds to human CD77, a small neutral glycosphingolipid also referred to as globotriaosylceramide (Gb3), Burkitt 's lymphoma associated antigen (BLA), ceramide trihexoside (CTH), and Pk blood group antigen. CD77 is expressed on a subset of germinal center B-lymphocytes including centroblasts that are found with greater frequency in tonsil and mucosal lymphoid tissues and less frequently in peripheral blood, bone marrow and spleen. It has been described as a receptor for Shiga Toxin, produced by Shigella dysenteriae, and Verotoxin 1 (VT1), produced by some strains of E. coli. CD19 has been reported as a ligand for CD77. This interaction may function in adhesion and signal transduction at a specific stage of B cell differentiation. Studies have shown that cross-linking CD77 induces apoptosis in some B cell lines.

Specifications
CD77 | |
Monoclonal | |
Brilliant Violet 510 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Initially Adherent Human Peripheral Blood Cells | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
5B5 | |
BLA; Gb3; Pk blood group antigen; CTH | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.