Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD77 Mouse anti-Human, Alexa Fluor 647, Clone: 5B5, BD

Catalog No. BDB563632 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563632 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563632 Supplier BD Biosciences Supplier No. 563632
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 5B5 monoclonal antibody specifically binds to human CD77, a small neutral glycosphingolipid also referred to as globotriaosylceramide (Gb3), Burkitt 's lymphoma associated antigen (BLA), ceramide trihexoside (CTH), and Pk blood group antigen. CD77 is expressed on a subset of germinal center B-lymphocytes including centroblasts that are found with greater frequency in tonsil and mucosal lymphoid tissues and less frequently in peripheral blood, bone marrow and spleen. It has been described as a receptor for Shiga Toxin, produced by Shigella dysenteriae, and Verotoxin 1 (VT1), produced by some strains of E. coli. CD19 has been reported as a ligand for CD77. This interaction may function in adhesion and signal transduction at a specific stage of B cell differentiation. Studies have shown that cross-linking CD77 induces apoptosis in some B cell lines.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD77
Applications Flow Cytometry
Classification Monoclonal
Clone 5B5
Conjugate Alexa Fluor 647
Description BLA; Gb3; Pk blood group antigen; CTH
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Initially Adherent Human Peripheral Blood Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.