Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79α Mouse anti-Human, Phycoerythrin, Clone: HM57, MilliporeSigma™

Catalog No. FCMAB208PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB208PMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB208PMI Supplier MilliporeSigma Supplier No. FCMAB208P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Milli-Mark™ Anti-CD79αcy-PE Antibody, clone HM57 is an antibody against CD79αcy for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79α
Applications Flow Cytometry
Classification Monoclonal
Clone HM57
Conjugate PE
Formulation Purified Mouse monoclonal IgG1 conjugated to PE in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.
Gene Accession No. NP_001774.1
Host Species Mouse
Immunogen Synthetic peptide sequence comprising amino acids 202-216 of the CD79α protein. This oligopeptide represents an intra-cytoplasmic C-terminal part of the protein.
Purification Method Protein A purfied
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.