Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79a Mouse anti-Human, PE, Clone: HM47, BD

Catalog No. BDB340579 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340579 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340579 Supplier BD Biosciences Supplier No. 340579
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD79a antibody, clone HM47, is derived from the hybridization of Sp2/0 mouse myeloma cells with spleen cells from BALB/c mice immunized with a synthetic peptide conjugated to thyroglobulin. The peptide has the sequence representing amino acids 202–216 inclusive as deduced from the human mb-1 cDNA sequence.

The CD79a antibody recognizes the cytoplasmic domain of a 47-kilodalton (kDa) membrane glycoprotein present on B lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79a
Applications Flow Cytometry
Classification Monoclonal
Clone HM47
Concentration 1.6μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human CD79a Peptide
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.