Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79a Mouse anti-Human, PE, Clone: HM57, BD

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 563777

Catalog No. BDB563777


Only null left
Add to Cart

Description

Description

The HM57 monoclonal antibody specifically binds to the cytoplasmic domain of CD79a. CD79a is a 47-kDa type 1 transmembrane glycoprotein present on B lymphocytes. CD79a is also referred to as mb-1, IGA and Ig-alpha (Ig- α ). It is expressed on B cells at various stages of differentiation, from the pre-B cell stage, probably befo re expression of cytoplasmic λ chain, to the plasma cell stage, in which it is detected only in the cytoplasm. CD79a associates with CD79b to form part of the B-cell receptor complex. It has been suggested that CD79a may play a role in mediating the transport of IgM to the cell surface. This antibody reportedly may crossreact with monkey, mouse, rat, bovine, canine, chicken, equine, guinea pig, porcine, and rabbit CD79a.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD79a
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CD79a Peptide
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
HM57
CD79A; CD79α; Immunoglobulin-associated alpha; Ig-alpha; IGA; Igα; MB-1
Mouse
Affinity Purified
RUO
Bovine, Canine, Chicken, Equine, Guinea Pig, Hamster, Human, Mouse, Pig, Rabbit, Rat
IgG1 κ
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.